missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BTLA Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3007
452.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
BTLA/CD272 Polyclonal specifically detects BTLA/CD272 in Mouse samples. It is validated for Western Blot.
Specifications
BTLA/CD272 | |
Polyclonal | |
Unconjugated | |
Q32MV9 | |
BTLA | |
Mouse myeloma cell line NS0-derived recombinant mouse BTLA Glu30-Gly176 (Pro41Glu, Thr45Asn, Thr47Lys, Gln52His, Arg55Trp, Gln63Glu, Cys85Trp) Accession # Q32MV9 | |
100 μg | |
Primary | |
Detects mouse BTLA in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human BTLA is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
Goat | |
Affinity Purified | |
RUO | |
151888 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse BTLA Antibody, R&D Systems™ > 100μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction