Learn More
R&D Systems™ Mouse FGFR4 (NP_032037) VersaClone cDNA
Fibroblast growth factor receptor 4
Brand: R&D Systems™ RDC0941
This item is not returnable.
View return policy
Description
The Mouse FGF R4 (NP_032037) VersaClone cDNA clone contains the complete ORF for FGFR4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The FGFR4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGF R4, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
R&D Systems™ Mouse FGFR4 (NP_032037) VersaClone cDNA
For Reasearch Use Only; Not for Use in Humans
Spot an opportunity for improvement?Share a Content Correction