Learn More
R&D Systems™ Mouse STING/TMEM173 (NP_082537) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC1469
Encompass
Product Code. 15771455
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Mouse STING/TMEM173 (NP_082537) VersaClone cDNA clone contains the complete ORF for STING/TMEM173, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The STING/TMEM173 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
STING/TMEM173
10 μg at 0.2 μg/μL
1150bp
endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173
Mouse
NP_082537
cDNA is provided in 10mM Tris-Cl, pH 8.142
340061
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Documents
One moment while we fetch your results.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
R&D Systems™ Mouse STING/TMEM173 (NP_082537) VersaClone cDNA
For Reasearch Use Only. Not for use in Humans.