Learn More
MP Biomedicals™ Casein
Low Trace Metals
Brand:  MP Biomedicals™ 0296012805
 View more versions of this product
Product Code. 11482831
  • 188.00 EUR / 5lb
Description
Precipitated from bovine milk by hydrochloric acid rather than lactic acid to remove most of the trace metals usually present in casein. Used in different animal diets.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Animal Diet
Powder
2.26 kg
15°C to 30°C
Protein 95%, Moisture 5%, Fat 1.5%, Lactose 0.2%, Ash 2.1%, pH (10% Solution) 4.6 to 5.1, Free Acids 0.2%
Used in different animal diet, blocking agen in immunochemistry
Rat
Product Title
MP Biomedicals™ Casein > Casein, 5 lbs