Learn More
MP Biomedicals Choline Control Diet, Modified
Brand: MP Biomedicals 0296041420
This item is not returnable.
View return policy
Description
Specifications
Choline Control Diet | |
Pellet | |
Powder | |
15°C to 30°C |
Vitamin Free Casein 10%, a-Protein 10%, Sucrose 51%, Alphacel, Non-Nutritive Bulk 5%, Lard 20%, Manganese Sulfates.H2O 150mg/kg diet, Zinc Chloride 20mg/kg diet, Chromium Potassium Sulfate.12H2O 55mg/kg diet, Sodium Selenite 1mg/kg diet, Salt Mixture Wesson 4% | |
To study liver injury in conjunction with MP's Methionine/Choline deficient diet | |
20,000 g |