missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Choline Control Diet, Modified

616.00 EUR - 2530.00 EUR

Specifications

Product Type Choline Control Diet
Components Vitamin Free Casein 10%, a-Protein 10%, Sucrose 51%, Alphacel, Non-Nutritive Bulk 5%, Lard 20%, Manganese Sulfates.H2O 150mg/kg diet, Zinc Chloride 20mg/kg diet, Chromium Potassium Sulfate.12H2O 55mg/kg diet, Sodium Selenite 1mg/kg diet, Salt Mixture Wesson 4%
Format Pellet
For Use With (Application) To study liver injury in conjunction with MP's Methionine/Choline deficient diet
Storage Requirements 15°C to 30°C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11491482
View Documents
MP Biomedicals
0296041450
50,000 g
2530.00 EUR
50kg
Please to purchase this item. Need a web account? Register with us today!
11471482
View Documents
MP Biomedicals
0296041410
10,000 g
616.00 EUR
10kg
Please to purchase this item. Need a web account? Register with us today!
11481482
View Documents
MP Biomedicals
0296041420
20,000 g
1150.00 EUR
20kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Choline Control Diet
Pellet
15°C to 30°C
Vitamin Free Casein 10%, a-Protein 10%, Sucrose 51%, Alphacel, Non-Nutritive Bulk 5%, Lard 20%, Manganese Sulfates.H2O 150mg/kg diet, Zinc Chloride 20mg/kg diet, Chromium Potassium Sulfate.12H2O 55mg/kg diet, Sodium Selenite 1mg/kg diet, Salt Mixture Wesson 4%
To study liver injury in conjunction with MP's Methionine/Choline deficient diet
Videos
SDS
Documents

Documents

Product Certifications