Learn More
MP Biomedicals™ β-Glucosidase
From Sweet Almonds
Brand: MP Biomedicals™ 0219519705
Description
Activity: >1000U/mg material. Unit Definition: One unit is the amount of enzyme which liberates 1.0μg of glucose per minute at 35°C, pH 5.0. Salt-free. Lyophilized.

Specifications
≥1000 U/mg | |
Synthesis of glucosides and fucosides | |
Sweet almonds | |
beta-Glucosidase |
White | |
-20°C | |
5 kU | |
Powder |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- beta-Glucosidase
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : beta-glucosidase
Your input is important to us. Please complete this form to provide feedback related to the content on this product.