missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ β-Glucosidase
From Sweet Almonds
Brand: MP Biomedicals™ 0219519780
View more versions of this product
This item is not returnable.
View return policy
Description
Activity: >1000U/mg material. Unit Definition: One unit is the amount of enzyme which liberates 1.0μg of glucose per minute at 35°C, pH 5.0. Salt-free. Lyophilized.

Specifications
≥1000 U/mg | |
Synthesis of glucosides and fucosides | |
Sweet almonds | |
beta-Glucosidase |
White | |
-20°C | |
100 kU | |
Powder |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ β-Glucosidase > 100KU
Spot an opportunity for improvement?Share a Content Correction