Learn More
MP Biomedicals™ LB Medium, Lennox, Capsules
Formulations for the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter; Lennox LB Medium also contains 5g NaCl per liter
Brand:  MP Biomedicals™ 113002141
 View more versions of this product
Product Code. 11333249
  • 1215.00 EUR / 5kg
Explore more special offers
Specifications
Molecular Biology Certified™ FDA
Capsule
Tryptone 10g/L, Yeast extract 5g/L, NaCl 5g/L
5000 g
15°C to 30°C
Microbiology Media
For growth of bacteria
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ LB Medium, Lennox, Capsules > Capsules; 5kg