Learn More
MP Biomedicals™ LB Medium, Lennox, Capsules
Formulations for the E. coli growth media known as LB Medium typically contain 10g of tryptone and 5g of yeast extract per liter; Lennox LB Medium also contains 5g NaCl per liter
Brand: MP Biomedicals™ 113002141
This item is not returnable.
View return policy
Description
Specifications
Molecular Biology Certified™ FDA | |
Capsule | |
Tryptone 10g/L, Yeast extract 5g/L, NaCl 5g/L | |
5000 g |
15°C to 30°C | |
Microbiology Media | |
For growth of bacteria |
Product Content Correction
Product Title
MP Biomedicals™ LB Medium, Lennox, Capsules > Capsules; 5kg
Spot an opportunity for improvement?Share a Content Correction