Learn More
MP Biomedicals™ Other Growth Factors and Supplements
Suitable for supplementing cell cultures and investigating various metabolic processes
Brand:  MP Biomedicals™ 0215232301
 View more versions of this product
Product Code. 11452622
  • 57.45 EUR / 1µg
Explore more special offers
Beschreibung
Beschreibung
†With streptomycin sulfate.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
-20°C
1mL of a sterile solution of 0.05M Na2HPO4, pH 7.5, albumin 0.5
>95%
Bovine Pituitary Glands
Growth Factor
Supplementing cell cultures and investigating various metabolic processes
1μg
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen
Sonderangebote
Sonderangebote