missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Other Growth Factors and Supplements
Suitable for supplementing cell cultures and investigating various metabolic processes
57.45 EUR
Specifications
For Use With (Application) | Supplementing cell cultures and investigating various metabolic processes |
---|
Product Code | Brand | Source | Product Type | Quantity | Content And Storage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Source | Product Type | Quantity | Content And Storage | Price | Quantity & Availability | ||
11452622
|
MP Biomedicals™
0215232301 |
Bovine Pituitary Glands | Growth Factor | 1μg | -20°C |
57.45 EUR
1µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Specifications
Supplementing cell cultures and investigating various metabolic processes |
Safety and Handling
missing translation for 'shelfLife' : 1 to 2 Weeks