missing translation for 'onlineSavingsMsg'
Learn More

MP7, MedChemExpress

MedChemExpress MP7 (PDK1 inhibitor) is a phosphoinositide-dependent kinase-1 (PDK1) inhibitor.

Encompass_Preferred

180.90 EUR - 1674.00 EUR

Specifications

Chemical Name or Material MP7
CAS 1001409-50-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H22F2N4O4
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16476068
Encompass_Preferred
View Documents
Medchem Express
HY-14440
10 mg
292.95 EUR
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16466466
Encompass_Preferred
View Documents
Medchem Express
HY-14440
100 mg
1674.00 EUR
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16462207
Encompass_Preferred
View Documents
Medchem Express
HY-14440
50 mg
1031.40 EUR
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16438677
Encompass_Preferred
View Documents
Medchem Express
HY-14440
5 mg
180.90 EUR
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MP7 (PDK1 inhibitor) is a phosphoinositide-dependent kinase-1 (PDK1) inhibitor.
Specifications

Specifications

MP7
Yellow
C28H22F2N4O4
DMSO : ≥ 100 mg/mL (193.61 mM)
516.5
99.83%
Solid
1001409-50-2
Research
PDK1 inhibitor
O=C(C1=CC=CN(CC2=CC=C(F)C(F)=C2)C1=O)N[C@H](C3=CC=CC=C3)COC4=CC=C(N5)C(NC5=O)=C4
516.5
Research
Cancer-Kinase/protease
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MP7, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.