missing translation for 'onlineSavingsMsg'
Learn More
Mucin, MP Biomedicals™ Product Code.: 11454931

Mucin, MP Biomedicals™

Product Code. 11454931 Shop All MP Biomedicals Products
500 mg
Click to view available options
Quantity:
100 mg
500 mg
1 g
This item is not returnable. View return policy

Product Code. 11454931

Brand: MP Biomedicals 0215574290

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 84195-52-8

Specifications

Color Tan
Quantity 500 mg
Formula Weight ≈400,0002
Physical Form Flaky Powder
Chemical Name or Material Mucin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mucin, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.