missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mucin, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
1 g
Specifications
Specifications
Color | Tan |
Quantity | 1 g |
Formula Weight | ≈400,0002 |
Physical Form | Flaky Powder |
Chemical Name or Material | Mucin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mucin, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction