Learn More
Axygen™ MultiRack Reload Tip System
MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.
41.53 EUR - 42.85 EUR
Specifications
Filtered Non-filtered
Sterility Non-sterile
Autoclavable Autoclavable
Certifications/Compliance ISO 13845
Clean Claims Free of detectable DNAse/RNAse, human DNA, and PCR inhibition
View More Specs
Products 4
Product Code Brand Volume (Metric) Tip Style Rack Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Tip Style Rack Color Price Quantity & Availability  
16302227
View Documents Promotion Details
Axygen™
MR-10XT-RL
10 μL Extended Length Red
41.53 EUR
Pack of 960
Estimated Shipment: 18-08-2025
to see stock.
Add to basket
16372227
View Documents Promotion Details
Axygen™
MR-200-RL
200 μL Universal Yellow
41.53 EUR
Pack of 960
Estimated Shipment: 18-08-2025
to see stock.
Add to basket
16332237
View Documents Promotion Details
Axygen™
MR-300-RL
300 μL Universal Green
42.85 EUR
Pack of 960
Estimated Shipment: 18-08-2025
to see stock.
Add to basket
16352217
View Documents Promotion Details
Axygen™
MR-1000XT-RL
1250 μL Extended Length Blue
41.53 EUR
Pack of 768
Estimated Shipment: 18-08-2025
to see stock.
Add to basket
Description
Description
The Axygen MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter, and reload packaging styles to meet the variety of needs in the lab setting. The MultiRack tip system offers standard and extended length tips covering volumes from 1 μL to 1250 μL. For racked and reload packaging styles, the tips are pre-loaded into easy-load color-coded deck inserts. With the smaller footprint and reusable rack, this system uses 30% less plastic, minimizing the environmental impact. The stackable design saves critical bench space. Axygen MultiRack Tip System products are made in the USA and meet ISO standards. All tips are free of detectable RNase, DNase, DNA, and pyrogens. For more details visit our Certificates page at www.corning.com/lifesciences.
Features
• Reusable rack with reload systems 
• Smaller footprint and stackable design, saving valuable bench space
• Extended tips offered in 10, 300, and 1250 μL (1000 μL extended length) volumes
• Color-coded racks for easy volume identification
Specifications
Specifications
Non-filtered
Autoclavable
Free of detectable DNAse/RNAse, human DNA, and PCR inhibition
Reload System
Yes
Polyethylene
Non-sterile
ISO 13845
Refill
Single-use
MultiRack
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Axygen™ MultiRack Reload Tip System