missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Mycophenolic acid, 98% Product Code.: 15562363
compliance-icons compliance-icons compliance-icons

Thermo Scientific Chemicals Mycophenolic acid, 98%

Product Code. 15562363
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15562363

Brand: Thermo Scientific Chemicals 459380010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24280-93-1
Molecular Formula C17H20O6
Molecular Weight (g/mol) 320.34
InChI Key HPNSFSBZBAHARI-RUDMXATFSA-N
Synonym mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid
PubChem CID 446541
ChEBI CHEBI:168396
IUPAC Name (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC

Specifications

Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 320.34
Percent Purity 98%
Chemical Name or Material Mycophenolic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466003 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog Number459380250,459380050,459380010
Lot NumberA0461767 Certificate TypeCertificate of Analysis Date24/04/2024 Catalog Number459380250,459380050,459380010
Lot NumberA0457200 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number459380250,459380050,459380010
Lot NumberA0448487 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number459380250,459380050,459380010
Lot NumberA0438842 Certificate TypeCertificate of Analysis Date15/02/2022 Catalog Number459380250,459380050,459380010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mycophenolic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360D-May damage the unborn child.
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Mycophenolic acid, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.