missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Phenylethyl)thiourea, 97%, Thermo Scientific™

29.55 EUR - 109.00 EUR

Chemical Identifiers

CAS 6815-00-5
Molecular Formula C9H12N2S
Molecular Weight (g/mol) 180.269
MDL Number MFCD00041194
InChI Key OFVGPHQYOCKLLM-UHFFFAOYSA-N
Synonym phenethylthiourea, 2-phenylethyl thiourea, n-phenethylthiourea, n-2-phenylethyl thiourea, phenethyl-thiourea, 1-phenethylthiourea, n-2-phenylethyl thioformamide, 1-2-phenylethyl thiourea, amino 2-phenylethyl amino methane-1-thione, 3-phenethylthiourea
PubChem CID 735845
IUPAC Name 2-phenylethylthiourea
SMILES C1=CC=C(C=C1)CCNC(=S)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11308485
SDS View Documents
Thermo Scientific Alfa Aesar
L10520.03
1 g N/A N/A N/A
11318485
SDS View Documents
Thermo Scientific Alfa Aesar
L10520.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(2-Phenylethyl)thiourea, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.