missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-valine, 96%, Thermo Scientific™

Product Code. 15408685
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 250714-85-3
Molecular Formula C12H14F3NO4S
Molecular Weight (g/mol) 325.30
MDL Number MFCD00173010
InChI Key KVNFXYSAEPLUHG-UHFFFAOYSA-N
Synonym 3-methyl-2-3-trifluoromethyl benzenesulfonamido butanoic acid, 3-trifluoromethyl phenyl sulfonyl valine, 3-methyl-2-3-trifluoromethyl phenyl sulfonyl amino butanoic acid, 3-methyl-2-3-trifluoromethyl phenyl-sulfonyl amino butanoic acid, valine,n-3-trifluoromethyl phenyl sulfonyl, 3-methyl-2-3-trifluoromethyl phenylsulfonamido butanoic acid, 3-methyl-2-3-trifluoromethyl phenyl sulfonylamino butanoic acid, n-3-trifluoromethyl phenyl sulfonyl valine
PubChem CID 2775542
IUPAC Name 3-methyl-2-[[3-(trifluoromethyl)phenyl]sulfonylamino]butanoic acid
SMILES CC(C)C(NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F)C(O)=O

Specifications

Melting Point 133°C to 134°C
Quantity 1 g
Formula Weight 325.31
Percent Purity 96%
Chemical Name or Material N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-valine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-valine, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.