missing translation for 'onlineSavingsMsg'
Learn More
N-Acetyl-L-cysteine (Crystalline), MP Biomedicals Product Code.: 11475012

N-Acetyl-L-cysteine (Crystalline), MP Biomedicals

Product Code. 11475012 Shop All MP Biomedicals Products
25 g
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
This item is not returnable. View return policy

Product Code. 11475012

Brand: MP Biomedicals 0219460325

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.191
MDL Number MFCD00004880
InChI Key PWKSKIMOESPYIA-BYPYZUCNSA-N
Synonym n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PubChem CID 12035
ChEBI CHEBI:28939
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)NC(CS)C(=O)O

Specifications

Color White
Alpha Vector Acetyl-L-cysteine
Quantity 25 g
Formula Weight 163.2
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Assay ≥96%
Chemical Name or Material N-Acetyl-L-cysteine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Acetyl-L-cysteine (Crystalline), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.