missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-α-Benzoyl-L-Argininamide Hydrochloride, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
Specifications
Specifications
CAS | 4299-03-0 |
Color | White |
Assay Percent Range | 99+% |
Molecular Formula | C13H20ClN5O2 |
Quantity | 5 g |
Synonym | s-n-1-amino-5-guanidino-1-oxopentan-2-yl benzamide hydrochloride, n-alpha-benzoyl-l-argininamide hydrochloride, 2s-5-carbamimidamido-2-phenylformamido pentanamide hydrochloride, bz-arg-nh hcl, bz-arg-nh2 hcl, benzoyl-l-arginine amide hydrochloride, n-benzoyl-l-arginine amide hydrochloride, benzoyl-arginine amide monohydrochloride monihydrate, nalpha-benzoyl-l-argininamidehydrochloridemonohydrate, 2s-5-diaminomethylidene amino-2-phenylformamido pentanamide hydrochloride |
InChI Key | PYZACNCNNFUUDO-PPHPATTJSA-N |
SMILES | C1=CC=C(C=C1)C(=O)NC(CCCN=C(N)N)C(=O)N.Cl |
IUPAC Name | N-[(2S)-1-amino-5-(diaminomethylideneamino)-1-oxopentan-2-yl]benzamide;hydrochloride |
Molecular Weight (g/mol) | 313.786 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-α-Benzoyl-L-Argininamide Hydrochloride, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction