missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Pyrrolidine, 97%, Thermo Scientific Chemicals

18.38 EUR - 238.19 EUR

Chemical Identifiers

CAS 86953-79-9
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00216581
InChI Key LPQZERIRKRYGGM-UHFFFAOYSA-N
Synonym 1-boc-pyrrolidine, n-boc-pyrrolidine, pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 1-pyrrolidinecarboxylate, 1-tert-butoxycarbonyl pyrrolidine, 1-pyrrolidinecarboxylic acid tert-butyl ester, boc-pyrrolidine, acmc-20aivt, pubchem9422, 1-boc-3-hydropyrrolidineShow MoreShow Less
PubChem CID 643455
IUPAC Name tert-butyl pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10264864
SDS View Documents
Thermo Scientific Acros
425820010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10366254
SDS View Documents
Thermo Scientific Acros
425820050
5 g Glass bottle
72.26 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
10613025
SDS View Documents
Thermo Scientific Acros
425820250
25 g Glass bottle
238.19 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-BOC-Pyrrolidine, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.