missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)aminoacetonitrile, 99%, Thermo Scientific Chemicals

66.53 EUR - 222.11 EUR

Chemical Identifiers

CAS 70591-20-7
Molecular Formula C15H12N2
Molecular Weight (g/mol) 220.28
MDL Number MFCD00009970
InChI Key VRLJFRODHVSTIK-UHFFFAOYSA-N
Synonym n-diphenylmethylene aminoacetonitrile, 2-diphenylmethylene amino acetonitrile, benzhydrylidene-amino-acetonitrile, 2-benzhydrylideneamino acetonitrile, 2-diphenylmethylidene amino acetonitrile, acetonitrile, diphenylmethylene amino, 4,4-diphenyl-3-azabut-3-enenitrile, pubchem13883, benzhydrylideneaminoacetonitrile
PubChem CID 612957
IUPAC Name 2-(benzhydrylideneamino)acetonitrile
SMILES N#CCN=C(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10214840
SDS View Documents
Thermo Scientific Acros
124140050
5 g Glass bottle
66.53 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
10637484
SDS View Documents
Thermo Scientific Acros
124140250
25 g Glass bottle
222.11 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-(Diphenylmethylene)aminoacetonitrile

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(Diphenylmethylene)aminoacetonitrile, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.