missing translation for 'onlineSavingsMsg'
Learn More
N-Fluorobenzenesulfonimide, 97%, Thermo Scientific Chemicals Product Code.: 11318855

N-Fluorobenzenesulfonimide, 97%, Thermo Scientific Chemicals

Product Code. 11318855
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11318855

Brand: Thermo Scientific Alfa Aesar L13955.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-Fluorobenzenesulfonimide fluorinates aromatics, enolates, azaenolates and carbanions in high yield. It is used for diastereoselective fluorination of Li enolates of chiral carboximides, for directed fluorination of ortho-lithiated aromatics and for a review of electrophilic N-F fluorinating agents. Asymmteric fluorinations is effected in the presence of a Pd-BINAP catalyst system at room temperature in an ionic liquid. It is a reagent employed in a palladium-catalyzed enantioselective fluorination of t-butoxycarbonyl lactones and lactams. It is also used in the electrophilic difluorination of dihalopyridines with butyl lithium and in the direct conversion of alcohols to dibenzenesulfonamides with triphenylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fluorobenzenesulfonimide fluorinates aromatics, enolates, azaenolates and carbanions in high yield. It is used for diastereoselective fluorination of Li enolates of chiral carboximides, for directed fluorination of ortho-lithiated aromatics and for a review of electrophilic N-F fluorinating agents. Asymmteric fluorinations is effected in the presence of a Pd-BINAP catalyst system at room temperature in an ionic liquid. It is a reagent employed in a palladium-catalyzed enantioselective fluorination of t-butoxycarbonyl lactones and lactams. It is also used in the electrophilic difluorination of dihalopyridines with butyl lithium and in the direct conversion of alcohols to dibenzenesulfonamides with triphenylphosphine.

Solubility
Very soluble in acetonitrile, dichloromethane or THF and less soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133745-75-2
Molecular Formula C12H10FNO4S2
Molecular Weight (g/mol) 315.333
MDL Number MFCD00144885
InChI Key RLKHFSNWQCZBDC-UHFFFAOYSA-N
Synonym n-fluorobenzenesulfonimide, nfsi, n-fluoro-n-phenylsulfonyl benzenesulfonamide, n-fluorodibenzenesulfonimide, n-fluorobenzensulfonimide, n-fluorobenzenesulphonimide, accufluor nfsi, n-fluorodi benzenesulfonyl amine, bis phenylsulfonyl fluoroamine, n-fluorobis phenylsulfonyl amine
PubChem CID 588007
IUPAC Name N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N(F)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 114°C to 118°C
Quantity 5 g
Beilstein 5348902
Solubility Information Very soluble in acetonitrile,dichloromethane or THF and less soluble in toluene.
Formula Weight 315.34
Percent Purity 97%
Chemical Name or Material N-Fluorobenzenesulfonimide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ08K010 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog NumberL13955.03,L13955.14,L13955.06
Lot Number10190556 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog NumberL13955.03,L13955.14,L13955.06
Lot NumberY01J004 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog NumberL13955.03,L13955.14,L13955.06
Lot NumberY08H037 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL13955.03,L13955.14,L13955.06
Lot NumberD08Z037 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13955.03,L13955.14,L13955.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Fluorobenzenesulfonimide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.