missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Hexane, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 mL
25 L
Packaging:
Metal Can
Plastic-coated Glass Bottle
Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Boiling Point | 69°C |
Quantity | 25 L |
Packaging | Metal Can |
Physical Form | Liquid |
Vapor Pressure | 160mbar at 20°C |
Viscosity | 0.31 mPaS at 20°C |
Formula Weight | 86.18g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Hexane, Certified AR for Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction