Learn More
N-Methyl-D-aspartic acid, 98+%, Thermo Scientific Chemicals
85.02 EUR - 321.60 EUR
Chemical Identifiers
CAS | 6384-92-5 |
---|---|
Molecular Formula | C5H9NO4 |
Molecular Weight (g/mol) | 147.13 |
MDL Number | MFCD00004226 |
InChI Key | HOKKHZGPKSLGJE-GSVOUGTGSA-N |
Synonym | n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartateShow More |
PubChem CID | 22880 |
ChEBI | CHEBI:31882 |
IUPAC Name | (2R)-2-(methylamino)butanedioic acid |
SMILES | CNC(CC(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463969
|
Thermo Scientific Alfa Aesar
J61361.MF |
50 mg |
85.02 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15443969
|
Thermo Scientific Alfa Aesar
J61361.MC |
100 mg |
122.61 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15453969
|
Thermo Scientific Alfa Aesar
J61361.ME |
500 mg | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6384-92-5 | |
147.13 | |
HOKKHZGPKSLGJE-GSVOUGTGSA-N | |
22880 | |
(2R)-2-(methylamino)butanedioic acid |
C5H9NO4 | |
MFCD00004226 | |
n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartateShow More | |
CHEBI:31882 | |
CNC(CC(=O)O)C(=O)O |
Specifications
187°C to 192°C | |
White | |
MFCD00004226 | |
14,6662 | |
HOKKHZGPKSLGJE-GSVOUGTGSA-N | |
(2R)-2-(methylamino)butanedioic acid | |
22880 | |
147.13 | |
Hygroscopic | |
Powder |
6384-92-5 | |
C5H9NO4 | |
1724431 | |
n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate | |
CNC(CC(=O)O)C(=O)O | |
147.13 | |
CHEBI:31882 | |
−15° (c=2 in Water) | |
≥98% | |
N-Methyl-D-aspartic acid |
Safety and Handling
missing translation for 'rtecsNumber' : CI9457000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.