missing translation for 'onlineSavingsMsg'
Learn More
n-Octyl butyrate, 97%, Thermo Scientific Chemicals Product Code.: 11401557

n-Octyl butyrate, 97%, Thermo Scientific Chemicals

Product Code. 11401557
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11401557

Brand: Thermo Scientific Alfa Aesar B24978.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-39-4
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00048939
InChI Key PWLNAUNEAKQYLH-UHFFFAOYSA-N
Synonym octyl butyrate, butanoic acid, octyl ester, n-octyl butyrate, butyric acid, octyl ester, n-octyl n-butyrate, n-octyl butanoate, unii-5yeu4o369l, butyric acid, octyl ester 8ci, octyl butyrate natural, fema no. 2807
PubChem CID 61030
IUPAC Name octyl butanoate
SMILES CCCCCCCCOC(=O)CCC

Specifications

Quantity 250 g
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material n-Octyl butyrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Octyl butyrate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.