missing translation for 'onlineSavingsMsg'
Learn More
N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals Product Code.: 10106823

N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals

Product Code. 10106823
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10106823

Brand: Thermo Scientific Acros 374790250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37595-74-7
Molecular Formula C8H5F6NO4S2
Molecular Weight (g/mol) 357.25
MDL Number MFCD00000404
InChI Key DIOHEXPTUTVCNX-UHFFFAOYSA-N
Synonym n,n-bis trifluoromethylsulfonyl aniline, phenyl triflimide, n-phenyl-bis trifluoromethanesulfonimide, n-phenyltrifluoromethanesulfonimide, n,n-bis-trifluoromethanesulfonyl aniline, 1,1,1-trifluoro-n-phenyl-n-trifluoromethylsulfonyl methanesulfonamide, n-phenylbis trifluoromethanesulfonimide, n-phenylbis trifluoromethanesulphonimide, 1,1,1-trifluoro-n-phenyl-n-trifluoromethyl sulfonyl methanesulfonamide, n,n-bis trifluoromethanesulfonyl anilineShow MoreShow Less
PubChem CID 142176
IUPAC Name 1,1,1-trifluoro-N-phenyl-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C1=CC=C(C=C1)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 99.0°C to 105.0°C
Color White or Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5N(SO2CF3)2
Quantity 25 g
Merck Index 15,7427
Formula Weight 357.25
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-Phenylbis(trifluoromethanesulfonimide)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0433645 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number374790250,374790050,374791000
Lot NumberA0439320 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number374790250,374790050,374791000
Lot NumberA0406973 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number374790250,374790050,374791000
Lot NumberA0416901 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number374790250,374790050,374791000
Lot NumberA0406972 Certificate TypeCertificate of Analysis Date28/03/2022 Catalog Number374790250,374790050,374791000
compliance-icons
Product Identifier
  • N-Phenylbis(trifluoromethanesulfonimide)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Phenylbis(trifluoromethanesulfonimide), 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.