Learn More
Thermo Scientific™ CryoTube™ Holders
Increase the functionality of your cryogenic tubes with these benchtop tube and vial holders in a variety of compartment configurations with single-handed capping and decapping.
Brand: Thermo Scientific™ 376589
22.20 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Description
Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.
- Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
- Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping
Specifications
Interlocking | |
Vial Holder | |
Nunc™ | |
4 x 10 |
PPO | |
40 Cryogenic Tubes | |
1/Pk., 15/Cs. |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
184128 | Certificate of Compliance | 22/02/2025 | 376589 |
171459 | Certificate of Compliance | 03/12/2024 | 376589 |
172861 | Certificate of Compliance | 03/12/2024 | 376589 |
174764 | Certificate of Compliance | 03/12/2024 | 376589 |
167718 | Certificate of Compliance | 03/12/2024 | 376589 |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
New Offer Save Now - Multiple Manufacturers