Learn More
Nasco Whirl-Pak™ Dry Cellulose Sponge Probe Write-On Bag
Convenient sterile absorbent sponge sampler in a sterile Whirl-Pak bag.
Brand: Nasco B01475WA
This item is not returnable.
View return policy
Description
- 8 in. (20.3 cm) long bendable polypropylene handle allows maximum contact of the sponge with the sampling surface
- Helps with the collection of hard-to-reach surfaces, around corners, and irregularly shaped areas.
- Innovative twist-off head holds the cellulose sponge securely and is quick and easy to remove from the handle once sample collection is completed.
- Sterilized write-on bag.
Specifications
Dry Sponge Probe | |
Designed for surface sampling | |
20.3 cm |
Tube | |
8 in. | |
50/Pk. |
Product Content Correction
Product Title
Nasco Whirl-Pak™ Dry Cellulose Sponge Probe Write-On Bag > 8 in. length; Box of 50
Spot an opportunity for improvement?Share a Content Correction