Learn More
Abeba™ Nature 4055 Shoes
Leather covered cork footbed is ESD dissipative.
96.15 EUR - 99.05 EUR
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, FO SRC |
---|---|
Color | Black |
Footbed Material | Cork |
Material | Leather |
Sole Features | Slip-resistant |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15681358
|
Abeba™
4055/34 |
34 |
96.15 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
15691358
|
Abeba™
4055/35 |
35 |
96.15 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
15601368
|
Abeba™
4055/36 |
36 |
96.15 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
15611368
|
Abeba™
4055/37 |
37 |
96.15 EUR
1 pair |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15621368
|
Abeba™
4055/38 |
38 |
96.15 EUR
1 pair |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15631368
|
Abeba™
4055/39 |
39 |
99.05 EUR
1 pair |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15641368
|
Abeba™
4055/40 |
40 |
96.15 EUR
1 pair |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15651368
|
Abeba™
4055/42 |
42 |
96.15 EUR
1 pair |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15661368
|
Abeba™
4055/43 |
43 |
96.15 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
15671368
|
Abeba™
4055/44 |
44 |
96.15 EUR
1 pair |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15681368
|
Abeba™
4055/45 |
45 |
96.15 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
15691368
|
Abeba™
4055/46 |
46 |
96.15 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
15601378
|
Abeba™
4055/47 |
47 |
96.15 EUR
1 pair |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
15611378
|
Abeba™
4055/48 |
48 |
96.15 EUR
1 pair |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Black
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Adjustable instep area
- Buckle fastening
- Resistant to oil and petrol
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
Slip-resistant | |
4055 |
Black | |
Leather | |
Rubber |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Abeba™ Nature 4055 Shoes