Learn More
NFIC Monoclonal Antibody (1D6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA519452
338.68 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Peptide Sequence: TEDMEGGISS PVKKTEMDKS PFNSPSPQDS PRLSSFTQHH RPVIAVHSGI ARSPHPSSAL HFPTTSILPQ TASTYFPHTA IRYPPHLNPQ DPLKDLVSLA CDPASQQPGP
NFIC Monoclonal antibody specifically detects NFIC in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.
Specifications
NFIC | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P08651 | |
NFIC | |
NFIC (NP_005588, 314 a.a. ∽ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunocytochemistry, Western Blot | |
1D6 | |
Unconjugated | |
NFIC | |
1110019L22Rik; 1500041O16Rik; AA589446; AI746521; CCAAT-box-binding transcription factor; CTF; CTF5; KIAA1439; NF1-A; NF1-B; NF1-C; NF1-X; NFI; NF-I; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; NFIB; NFI-B; Nfic; NFI-C; NFIX; NFI-X; Nuclear factor 1 C-type; Nuclear factor 1/A; Nuclear factor 1/B; nuclear factor 1/C; Nuclear factor 1/X; nuclear factor 1-C2; nuclear factor I C; Nuclear factor I/A; Nuclear factor I/B; nuclear factor I/C; nuclear factor I/C (CCAAT-binding transcription factor); Nuclear factor I/X; olfactory epithelium nuclear factor I-C; TGGCA-binding protein | |
Mouse | |
Affinity chromatography | |
RUO | |
4782 | |
-20° C, Avoid Freeze/Thaw Cycles | |