missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Nicotinamide, 99% Product Code.: 10117953

Thermo Scientific Chemicals Nicotinamide, 99%

Product Code. 10117953
2.5 kg, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10117953

Brand: Thermo Scientific Chemicals 128270025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006395
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
Synonym nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide
PubChem CID 936
ChEBI CHEBI:17154
IUPAC Name pyridine-3-carboxamide
SMILES C1=CC(=CN=C1)C(=O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.0°C to 131.0°C
Color White
pH 6.0 to 7.5 (5% soln. at 20°C)
Boiling Point 150.0°C to 160.0°C
Flash Point >150°C
Loss on Drying 0.5% max. (4 hrs) (silica gel)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 22, 40
Merck Index 15, 6608
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: 1g/1.5mL alcohol,1g/10mL glycerol
Formula Weight 122.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Nicotinamide, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467572 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number128275000,128271000,128270025
Lot NumberA0462243 Certificate TypeCertificate of Analysis Date24/04/2024 Catalog Number128275000,128271000,128270025
Lot NumberA0457892 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number128275000,128271000,128270025
Lot NumberA0453917 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number128275000,128271000,128270025
Lot NumberA0451799 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number128275000,128271000,128270025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Nicotinamide, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.