missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIT2 Monoclonal Antibody (OTI3F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501098
This item is not returnable.
View return policy
Description
Has a omega-amidase activity. The role of omega-amidase is to remove potentially toxic intermediates by converting alpha-ketoglutaramate and alpha-ketosuccinamate to biologically useful alpha-ketoglutarate and oxaloacetate, respectively. Overexpression decreases the colony-forming capacity of cultured cells by arresting cells in the G2 phase of the cell cycle.Specifications
NIT2 | |
Monoclonal | |
0.72 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NQR4 | |
NIT2 | |
Full length human recombit protein of human NIT2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3F3 | |
Unconjugated | |
NIT2 | |
HEL-S-8a | |
Mouse | |
Affinity Chromatography | |
RUO | |
56954 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |