missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIT2 Monoclonal Antibody (OTI3F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00 EUR - 540.87 EUR
Specifications
Antigen | NIT2 |
---|---|
Clone | OTI3F3 |
Concentration | 0.72 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17533291
|
Origene Technologies
TA501098S |
30 μL |
242.00 EUR
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17513291
|
Origene Technologies
TA501098 |
100 μL |
540.87 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NIT2 Monoclonal antibody specifically detects NIT2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
NIT2 | |
0.72 mg/mL | |
Unconjugated | |
Mouse | |
Human | |
56954 | |
Primary |
OTI3F3 | |
Monoclonal | |
Liquid | |
RUO | |
Q9NQR4 | |
IgG2b | |
-20° C, Avoid Freeze/Thaw Cycles |