Learn More
Novus Biologicals™ Recombinant Human HIF-1 alpha Protein
Highly purified. Generating reliable and reproducible results.
Brand:  Novus Biologicals™ NBC1-18422
Encompass
Product Code. 15714106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
An un-tagged recombinant protein corresponding to the amino acids 530-826 of Human HIF-1 alpha The Recombinant Human HIF-1 alpha Protein is derived from E. coli. The Recombinant Human HIF-1 alpha Protein has been validated for the following applications: SDS-Page, Block/Neutralize.
Specifications
Specifications
Q16665
3091
HIF-1 alpha Protein
0.1mg
Hif-1a (530-826 aa) Human, Recombinant, E.coli (NP_001521).
RUO
Unlabeled
20mM Tris-HCl buffer (pH 7.5) 1mM DTT
32.8kDa
Protein
E. Coli
−80°C. Avoid freeze-thaw cycles.
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8
Yes
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human HIF-1 alpha Protein