Learn More
Tocris Bioscience™ NSC 624206
Ubiquitin-activating enzyme (E1) inhibitor
Brand: Tocris Bioscience™ 4993/10
This item is not returnable.
View return policy
Description
NSC 624206 is an ubiquitin-activating enzyme (E1) inhibitor; prevents tumour suppressor protein p27 ubiquitination in vitro. Blocks the ubiquitin-thioester formation step of the E1 activation reaction, but displays no effect on ubiquitin adenylation.Specifications
>98% | |
NSC 624206 | |
NSC 624206 |
10 mg | |
410.51 |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction