missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NSC 624206
Ubiquitin-activating enzyme (E1) inhibitor
277.00 EUR - 1070.00 EUR
Specifications
Purity | >98% |
---|---|
Product Type | NSC 624206 |
Molecular Weight (g/mol) | 410.51 |
Inhibitors | NSC 624206 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729957
|
Tocris Bioscience™
4993/10 |
10 mg |
277.00 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18780458
|
Tocris Bioscience™
4993/50 |
50 mg |
1070.00 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ubiquitin-activating enzyme (E1) inhibitor; prevents tumour suppressor protein p27 ubiquitination in vitro. Blocks the ubiquitin-thioester formation step of the E1 activation reaction, but displays no effect on ubiquitin adenylation.Specifications
>98% | |
NSC 624206 | |
410.51 | |
NSC 624206 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title