Learn More
Tocris Bioscience™ NSC 624206
Ubiquitin-activating enzyme (E1) inhibitor
Brand:  Tocris Bioscience™ 4993/50
 View more versions of this product
Product Code. 18780458
  • 1070.00 EUR / 50mg
[resources not loaded]
Explore more special offers
Description
Description
NSC 624206 is an ubiquitin-activating enzyme (E1) inhibitor; prevents tumour suppressor protein p27 ubiquitination in vitro. Blocks the ubiquitin-thioester formation step of the E1 activation reaction, but displays no effect on ubiquitin adenylation.
Specifications
Specifications
>98%
NSC 624206
NSC 624206
50 mg
410.51