Learn More
NSD2 Monoclonal Antibody (CL1063), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524621
370.68 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Immunogen sequence: SANGKTPSCE VNRECSVFLS KAQLSSSLQE GVMQKFNGHD ALPFIPADKL KDLTSRVFNG EPGAHDAKLR FESQEMKGIG TPPNTTPIKN GSPEIKLKIT KTYMNGKPLF ESSICGD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%. Binds to an epitope located within the peptide sequence ALPFIPADKL as determined by overlapping synthetic peptides.
This gene encodes a protein that contains four domains present in other developmental proteins: a PWWP domain, an HMG box, a SET domain, and a PHD-type zinc finger. It is expressed ubiquitously in early development. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene maps to the 165 kb WHS critical region and has also been involved in the chromosomal translocation t(4;14)(p16.3;q32.3) in multiple myelomas. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. Some transcript variants are nonsense-mediated mRNA (NMD) decay candidates, hence not represented as reference sequences.
Specifications
NSD2 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O96028 | |
NSD2 | |
Recombinant protein corresponding to Human NSD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
CL1063 | |
Unconjugated | |
NSD2 | |
5830445G22Rik; 9430010A17Rik; AW555663; C130020C13Rik; D030027O06Rik; D930023B08Rik; Histone-lysine N-methyltransferase NSD2; histone-lysine N-methyltransferase NSD2; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; Kiaa1090; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase NSD2; mKIAA1090; MMSET; multiple myeloma SET domain containing protein type III; multiple myeloma SET domain-containing protein; NSD2; nuclear receptor binding SET domain protein 2; nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; putative histone-lysine N-methyltransferase NSD2-like protein; REIIBP; RGD1565590; trithorax/ash1-related protein 5; TRX5; WHS; Whsc1; Whsc1l; Wolf-Hirschhorn syndrome candidate 1; Wolf-Hirschhorn syndrome candidate 1 (human); Wolf-Hirschhorn syndrome candidate 1 protein; Wolf-Hirschhorn syndrome candidate 1 protein homolog | |
Mouse | |
Protein A | |
RUO | |
7468 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only