Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Brand: Thermo Scientific™ 167525
38.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Connections Made Easy:- Plug a media bag or tubing set to any Cell Factory system for enhanced aseptic connectivity for filling and harvesting
- Save time and reduce costs
- Minimize cleaning processes such as autoclaving and associated validations
- Prevent build up of back pressure during filling application
- Get additional air venting with the venting options
Specifications
Filter Adaptor Cap, Sterile | |
Cell Factory Systems (All) | |
1 | |
Sterile |
HDPE | |
20 | |
Filter Adapter Cap, Sterile | |
Filter Adapter |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
167369 | Certificate of Compliance | 06/12/2024 | 167525 |
178336 | Certificate of Compliance | 03/12/2024 | 167525 |
175755 | Certificate of Compliance | 03/12/2024 | 167525 |
169239 | Certificate of Compliance | 03/12/2024 | 167525 |
177599 | Certificate of Compliance | 03/12/2024 | 167525 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.