Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.
Brand: Thermo Scientific™ 164327
364.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
- Ideal for adherent cells
- Boost production efficiency with more surface area in a small footprint
- Reduce contamination risks with a closed system
Specifications
Original Cell Factory | |
2/Cs. | |
Cell Factory System | |
Less waste, renewable material | |
Adapter Cap and Port Cover Cap | |
Nunclon Delta |
Polystyrene | |
1 Ea. | |
Nunclon™ Delta | |
Flat | |
6320 cm² | |
10 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1423036 | Certificate of Processing | 12/03/2025 | 164327 |
1413357 | Certificate of Processing | 19/12/2024 | 164327 |
1423036 | Certificate of Compliance | 28/02/2025 | 164327 |
1283542 | Certificate of Compliance | 11/12/2024 | 164327 |
183672 | Certificate of Compliance | 06/12/2024 | 164327 |
Accessories and Services
-
Thermo Scientific™ Nunc™ Cell Factory™ System A... START-UP Kit for Cell Factory System, St... Product Code: 10274451 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.