Learn More
Thermo Scientific™ NuPAGE™ Bis-Tris XCell SureLock™ Mini-Cell Welcome Pack, 4 to 12%
The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell.
644.00 EUR
Specifications
For Use With (Equipment) | XCell SureLock Mini-Cell |
---|---|
Gel Percentage | 4 to 12% |
Gel Size | Mini |
Gel Thickness | 1.0 mm |
Gel Type | Bis-Tris |
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
17739893
|
Thermo Scientific™
NP032D |
10-well |
644.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17749893
|
Thermo Scientific™
NP032E |
12-well |
644.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17759893
|
Thermo Scientific™
NP032F |
15-well |
644.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NuPAGE Bis-Tris Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.
The NuPAGE Bis-Tris Welcome Pack contains:
• XCell SureLock Mini-Cell (EI0001)
• NuPAGE Bis-Tris mini gels (2 boxes, 20 gels)
• NuPAGE MES SDS Running Buffer, 20X (NP0002)
• NuPAGE LDS Sample Buffer, 4X (NP0007)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)
About the XCell SureLock Mini-Cell
The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.
About NuPAGE Bis-Tris gels
Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use.
Transfer
For transferring your proteins to a membrane, we recommend using for NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).
Specifications
XCell SureLock Mini-Cell | |
Mini | |
Bis-Tris | |
NuPAGE™ | |
Approved for shipment at room temperature and wet ice |
4 to 12% | |
1.0 mm | |
3.5 to 260 kDa | |
1 Welcome Pk. |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.