Learn More
Cole Parmer Oakton Ion-Selective Electrode, Fluoride
Oakton Ion-Selective Electrode, Fluoride is fast and accurate and does not require an external reference - they're easier to use than separate sensing and reference half-cell electrodes.
Brand: Cole Parmer 27502-19
406.08 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Oakton Ion-Selective Electrode, Fluoride is fast and accurate and does not require an external reference - they're easier to use than separate sensing and reference half-cell electrodes.
- Electrodes are double-junction combination ion-selective electrodes and are convenient to use in places where two electrodes won't fit.
- Glass body provides high-quality measurements for laboratory applications.
- Improved glass seal enhances performance and reliability.
- Sealed inner junctions and refillable outer junctions accommodate special reference fill solutions - ideal for low-concentration measurements.
- Solid-state electrodes have twelve polishing strips.
- Use with a meter that has a resolution of 0.1 mV for the most accurate results.
Specifications
BNC | |
Glass | |
ISE |
Double-junction | |
30 mL bottle of Fluoride reference solution, Reference filling pipette, 1 m cable, BNC connector, and Instruction manual |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers