Learn More
Welch Ilmvac™ Oil Mist Filter
Eliminate the emission of oil mist. Welch Ilmvac™ Oil Mist Filters feature transparent polyamide housing that enables an easy check of the degree of contamination or saturation of the filter.
Brand: Welch Ilmvac™ 700011
Additional Details : Weight : 0.06300kg
Description
- Economical mist filters with KF flanges
- If the filter insert becomes blocked, an integrated pressure control valve opens automatically and reduces the built-up pressure immediately
- Worn out filter inserts can be replaced easily
Specifications
1 Pack | |
DN 25 KF | |
Oil Mist Filter |