Learn More
Welch Ilmvac™ Oil Mist Filter
Eliminate the emission of oil mist. Welch Ilmvac™ Oil Mist Filters feature transparent polyamide housing that enables an easy check of the degree of contamination or saturation of the filter.
133.56 EUR - 153.03 EUR
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10613682
|
Welch Ilmvac™
700010 |
DN 16 KF |
133.56 EUR
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
10330641
|
Welch Ilmvac™
700011 |
DN 25 KF |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Economical mist filters with KF flanges
- If the filter insert becomes blocked, an integrated pressure control valve opens automatically and reduces the built-up pressure immediately
- Worn out filter inserts can be replaced easily
Specifications
1 Pack | |
Oil Mist Filter |