Learn More
Ohaus™ OIML F1 Weight Set
Adjusted to Meet or Exceed OIML F1 Tolerances
Brand: Ohaus™ 80780412
This item is not returnable.
View return policy
Description
- Weights Sets in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free to protect your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
0.300 kg | |
Protective Case |
Stainless Steel, Aluminum | |
100 g to 1 mg | |
21/Pk. |