Learn More
Ohaus™ OIML F1 Weight Set
Adjusted to Meet or Exceed OIML F1 Tolerances
308.00 EUR - 2425.00 EUR
Specifications
Certifications/Compliance No Certificate
Includes Protective Case
View More Specs
Products 15
Product Code Brand Mass Price Quantity & Availability  
Product Code Brand Mass Price Quantity & Availability  
16672462
View Documents
Ohaus™
80780408
500 mg to 1 mg
308.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16682462
View Documents
Ohaus™
80780410
50 g to 1 mg
610.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16692462
View Documents
Ohaus™
80780411
50 g to 1 g
380.00 EUR
Each
In Stock
to see stock.
Add to basket
16602472
View Documents
Ohaus™
80780412
100 g to 1 mg
672.00 EUR
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
16612472
View Documents
Ohaus™
80780413
100 g to 1 g
457.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16622472
View Documents
Ohaus™
80780414
2 x 200 g to 1 mg
872.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16632472
View Documents
Ohaus™
80780415
2 x 200 g to 1 g
580.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16642472
View Documents
Ohaus™
80780418
500 g to 1 mg
928.00 EUR
Each
In Stock
to see stock.
Add to basket
16652472
View Documents
Ohaus™
80780419
500 g to 1 g
687.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16662472
View Documents
Ohaus™
80780420
1 kg to 1 mg
1530.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16672472
View Documents
Ohaus™
80780421
1 kg to 1 g
1315.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16682472
View Documents
Ohaus™
80780422
2 x 2 kg to 1 mg
1990.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16692472
View Documents
Ohaus™
80780423
2 x 2 kg to 1 g
1870.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16602482
View Documents
Ohaus™
80780426
5 kg to 1 mg
2425.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
16612482
View Documents
Ohaus™
80780427
5 kg to 1 g
2140.00 EUR
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
Description
Description
  • Weights Sets in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free to protect your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
Specifications
No Certificate
Protective Case