Learn More
Olopatadine hydrochloride, MedChemExpress
Product Code. 16418488
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16418488
Brand: Medchem Express HYB0426A10mg
MedChemExpress Olopatadine hydrochloride (ALO4943A) is a histamine blocker used to treat allergic conjunctivitis.
Olopatadine hydrochloride (ALO4943A) is a histamine blocker used to treat allergic conjunctivitis.
Specifications
Chemical Name or Material Olopatadine hydrochloride
CAS 140462-76-6
Color White
Purity Grade Notes Research
Molecular Formula C21H24ClNO3
Quantity 10 mg
Synonym ALO4943A KW4679
Solubility Information DMSO : 50 mg/mL (133.74 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (17.84 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=C(C/2=C1)OCC3=CC=CC=C3C2=C\CCN(C)C.Cl
Molecular Weight (g/mol) 373.87
Formula Weight 373.87
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation