Learn More
Omadacycline hydrochloride, MedChemExpress
MedChemExpress Omadacycline (PTK 0796) hydrochloride, a first-in-class orally active aminomethylcycline antibacterial, is a member of the tetracycline class of antibiotics.
Brand: Medchem Express HY-14865C
Description
Omadacycline hydrochloride acts through the inhibition of bacterial protein synthesis by binding to the 30S ribosomal subunit. Omadacycline hydrochloride possesses broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. Omadacycline hydrochloride can be used for the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.Specifications
Omadacycline hydrochloride | |
Dark Brown | |
C29H40N4O7.HCl | |
PTK0796 hydrochloride Amadacycline hydrochloride | |
O=C(C1=C(O)[C@@H](N(C)C)[C@@](C[C@@]2([H])C(C(C3=C(O)C(CNCC(C)(C)C)=CC(N(C)C)=C3C2)=O)=C4O)([H])[C@@]4(O)C1=O)N.Cl | |
593.11 | |
Research | |
COVID-19-immunoregulation |
1196800-39-1 | |
Research | |
50 mg | |
DMSO : 105 mg/mL (177.03 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (84.30 mM) | |
593.11 | |
98.02% | |
Solid |