Learn More
ONO-7300243, MedChemExpress
Product Code. 16463208
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16463208
Brand: Medchem Express HY100882
MedChemExpress ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.
ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.
Specifications
Chemical Name or Material ONO-7300243
CAS 638132-34-0
Color White
Purity Grade Notes Research
Molecular Formula C28H31NO5
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (216.66 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=C(CN(C(C2=CC(OC)=C(C)C(OC)=C2)=O)CCCC3=CC=CC=C3)C=C1
Molecular Weight (g/mol) 461.55
Formula Weight 461.55
Percent Purity 98.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation